Anti-CTNNB1 (phospho-Tyr333) Rabbit Polyclonal Antibody
Catalog # 77710-292
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CTNNB1 (phospho-Tyr333)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P35222
- Antigen Synonyms:b-catenin|CTNB1_HUMAN|OTTHUMP00000209288|CHBCAT|CATNB|Catenin beta 1|FLJ25606|OTTHUMP00000165222|OTTHUMP00000165223|beta 1|Beta-catenin|FLJ37923|Catenin beta-1|DKFZp686D02253|OTTHUMP00000162082|Beta catenin|CTNNB|Catenin (cadherin associated protein)|Cadherin associated protein|OTTHUMP00000209289|CTNNB1|88kDa
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around phosphorylation site of tyrosine 333 (Y-T-Y(p)-E-K) derived from Human β-catenin
- Purification:Antigen affinity purified
- Cat. No.:77710-292
- Packaging:Vial
Specifications
About this item
WB:1:500-1:1000, IHC:1:50-1:100
Type:
Antigen: CTNNB1 (phospho-Tyr333)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human