Anti-RAB6C Rabbit Polyclonal Antibody
100 µg, Vial
Catalog # 77700-424
Supplier: AFG Bioscience
: A15221-100UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
WB:1:500-1:2000, ELISA:1:40000
Type:
Antigen: RAB6C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77700-424
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:RAB6C
- Gene ID:Q9H0N0
- Immunogen:Synthesized peptide derived from the Internal region of Human Rab 6C
- Conjugation:Unconjugated
- Antigen Synonyms:RAB6C_HUMAN|Ras-related protein Rab-6C|Ras related protein Rab 6C|RAB 6C|Rab6-like protein WTH3|Rab6 like protein WTH3|WTH3|RAB6C|RAB6C member RAS oncogene family
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.



