Anti-TP53 Rabbit Polyclonal Antibody
100 µg, Vial
Catalog # 77700-265
Supplier: AFG Bioscience
Supplier #: A15020-100UG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: TP53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Size:100 µg
- Packaging:Vial
- Clonality:Polyclonal
- Purification:Antigen affinity purified
- Form:Liquid
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Isotype:IgG
- Host:Rabbit
- Cat. No.:77700-265
- Reactivity:Human|Rat|Mouse
- Western Blot:Yes
- Shipping Temperature:Shipped on blue ice
- Antigen Symbol:TP53
- Gene ID:P04637
- Immunogen:Synthesized peptide derived from Human p53 around the non-phosphorylation site of T18
- Conjugation:Unconjugated
- Antigen Synonyms:P53_HUMAN|Tp53|Tumor protein 53|Tumor suppressor p53|BCC7|Cellular tumor antigen p53|TRP53|tumor antigen p55|Tumor protein p53|Mutant tumor protein 53|LFS1|Antigen NY-CO-13|Phosphoprotein p53|Transformation related protein 53|FLJ92943|p53|p53 tumor suppressor
- ELISA:Yes
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.



