Anti-SRCIN1 Rabbit Polyclonal Antibody
Catalog # 77700-240
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:SRCIN1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:Q9C0H9
- Antigen Synonyms:p130Cas associated protein|SRCIN1|P140|SNAP-25-interacting protein|SNIP|SRCN1_HUMAN|p140Cap|p130Cas-associated protein|SRC kinase signaling inhibitor 1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the Internal region of Human p140Cap
- Purification:Antigen affinity purified
- Cat. No.:77700-240
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, ELISA:1:20000
Type:
Antigen: SRCIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat