Anti-OR5F1 Rabbit Polyclonal Antibody
Catalog # 77700-207
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR5F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:O95221
- Antigen Synonyms:Olfactory receptor OR11-167|Olfactory receptor 5F1|Olfactory receptor 11-10|OR11-10|OR5F1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 5F1
- Purification:Antigen affinity purified
- Cat. No.:77700-207
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: OR5F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human