Anti-OR10J1 Rabbit Polyclonal Antibody
Catalog # 77700-162
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:OR10J1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P30954/Q2M1M8/Q5VSV1
- Antigen Synonyms:Olfactory receptor 10J1
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human Olfactory receptor 10J1
- Purification:Antigen affinity purified
- Cat. No.:77700-162
- Packaging:Vial
Specifications
About this item
WB:1:500-1:2000, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: OR10J1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human