Anti-DNAJB1 Rabbit Polyclonal Antibody
77699-745
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Symbol:DNAJB1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:P25685
- Antigen Synonyms:HDJ 1|Human DnaJ protein 1|DnaJ protein homolog 1|DnaJ homolog subfamily B member 1|Radial spoke 16 homolog B|Heat shock protein 40|HDJ-1|DNAJ 1|RSPH16B|HDJ1|Dnajb1|DNAJB 1|DnaJ heat shock protein family (Hsp40) member B1|HSPF1|DNAJB1 protein|DnaJ (Hsp40) homolog subfmaily B member 1|DNAJ B1|DNJB1_HUMAN|Heat shock 40 kDa protein 1|HSPF 1|Heat shock 40kD protein 1|HSP40|DNAJ1|Sis1|Hsp 40
- Storage Buffer:PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from the C-terminal region of Human HSP40
- Purification:Antigen affinity purified
- Cat. No.:77699-745
- Packaging:Vial
WB:1:500-1:2000, IHC:1:100-1:300, IF:1:200-1:1000, ELISA:1:10000
Type:
Antigen: DNAJB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat