
Pre-Mixed GFAA matrix modifier 2: 1000 µg/mL Pd & 600 µg/mL Mg(NO3)2 (w/v) in nitric acid 2%, certified reference material, VHG
Catalog # 77697-697
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:250 ml
- Cat. No.:77697-697
Specifications
Specification Test Results
- Palladium (Pd):1000 ± 5 µg/mL
- Magnesium Nitrate [Mg(NO3)2]:600.0 ± 3.0 µg/mL
Specification Test Results
About this item
:
:GHS05
Storage Temperature:Ambient