
Nickel (II) nitrate 1% Ni(NO3)2 (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Catalog # 77697-695
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:77697-695
Specifications
Specification Test Results
- Nickel Nitrate Ni(NO3)2:10,000 ± 50 µg/m
Specification Test Results
About this item
:
:182.7
:GHS05
Storage Temperature:Ambient
CAS Number:13138-45-9
Formula:N₂NiO₆
MDL Number:MFCD00011139