
Magnesium nitrate 1% Mg(NO₃)₂ (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Catalog # 77697-694
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:77697-694
Specifications
Specification Test Results
- Magnesium Nitrate (Mg(NO3)2) :10,000 ± 50 µg/mL
Specification Test Results
About this item
:
:GHS05
CAS Number:10377-60-3
:13446-18-9
Formula:Mg(NO₃)₂