Ammonium phosphate 10% (w/v) in nitric acid 2% matrix modifier, for graphite furnace AAS (CRM), VHG
Catalog # 77693-168
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:100 ml
- Cat. No.:77693-168
Specifications
Specification Test Results
- Ammonium Phosphate (NH4H2PO4):100,000 ± 500 µg/mL
Specification Test Results
About this item
:
:115.03 g/mol
CAS Number:7722-76-1
Formula:NH₄H₂PO₄
MDL Number:MFCD00003396