Multi-Element 11 Component Standard: Al,Cu @5µg/mL; Mg @10µg/mL; Zn @20µg/mL; Fe,Pb @25µg/mL; Ca @50µg/mL; B @250µg/mL; P @1000µg/mL; K @2500µg/mL; Na @5000µg/mL in 3% HNO3 (v/v),VHG,LGC Standards
Catalog # 77692-990
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:3% HNO₃
- Description:Al, Cu @ 5µg/mL; Mg @ 10µg/mL; Zn @ 20µg/mL; Fe, Pb @ 25µg/mL; Ca @ 50µg/mL; B @ 250µg/mL; P @ 1000µg/mL; K @ 2500µg/mL; Na @ 5000µg/mL in 3% HNO3 (v/v), 500mL
- Size:500 ml
- Cat. No.:77692-990