Order Entry
Puerto Rico
ContactUsLinkComponent
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Catalog # 77692-993
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Catalog # 77692-993
Supplier:  LGC Standards
Supplier Number:  VHG-44CS2Z-100
New Product
CAS Number:  

Specifications

  • Matrix:
    20% HCl
  • Description:
    200.7 Rev. 4.4 - Calibration Standard 2: Sn @ 200; Li, Sb @ 500; Al, Fe, Mo, Na, SiO2, Ti @ 1000 µg/mL in 20% HCl, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77692-993

Specifications

About this item