200.7, 6010 - Quality Control (QCS) Solution 2: Al, Fe, K, Li, Mo, Na, Sb, SiO2, Sn, Ti @ 100 µg/mL in 20% HCl, tr. HF, VHG, LGC Standards
Catalog # 77692-996
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:20% HCl
- Description:200.7, 6010 - Quality Control (QCS) Solution 2: Al, Fe, K, Li, Mo, Na, Sb, SiO2, Sn, Ti @ 100 µg/mL in 20% HCl, tr. HF
- Size:100 ml
- Cat. No.:77692-996