Order Entry
Puerto Rico
ContactUsLinkComponent
Titanium AA Standard: Ti @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Titanium AA Standard: Ti @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77695-316
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Titanium AA Standard: Ti @ 1000 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77695-316
Supplier:  LGC Standards
Supplier Number:  VHG-AATINF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    Titanium AA Standard: Ti @ 1000 µg/mL in 5% HNO3, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77695-316

Specifications

About this item