Bismuth Internal Standard: Bi @ 0.793 g/L in Mineral Oil, VHG, LGC Standards
Catalog # 77693-044
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:0.793 g/L
- Matrix:Mineral oil
- Description:Bismuth Internal Standard: Bi @ 0.793 g/L in Mineral Oil
- Size:400 g
- Cat. No.:77693-044