Chlorine Standard: Cl @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
77693-056
New Product
:
- Concentration:10 µg/g
- Matrix:Mineral oil
- Description:Chlorine Standard: Cl @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77693-056