Chlorine Standard: Cl @ 100 µg/g (0.0100 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
77693-055
New Product
:
- Concentration:100 µg/g
- Matrix:Mineral oil
- Description:Chlorine Standard: Cl @ 100 µg/g (0.0100 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77693-055