Chlorine Standard: Cl @ 50 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77693-066
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:50 µg/g
- Matrix:Mineral oil
- Description:Chlorine Standard: Cl @ 50 µg/g in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77693-066