Order Entry
Puerto Rico
ContactUsLinkComponent
Chlorine Standard: Cl @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Chlorine Standard: Cl @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77693-065
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Chlorine Standard: Cl @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77693-065
Supplier:  LGC Standards
Supplier Number:  VHG-CLOIL-500-100
New Product
CAS Number:  

Specifications

  • Concentration:
    500 µg/g
  • Matrix:
    Mineral oil
  • Description:
    Chlorine Standard: Cl @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil
  • Size:
    100 ml
  • Cat. No.:
    77693-065

Specifications

About this item