Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chlorine Standard: Cl @ 5000 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
  77693-064
New Product
 :  
undefined
Chlorine Standard: Cl @ 5000 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
  77693-064
 :  VHG-CLOIL-5000-500
New Product
 :  

 

  • Concentration:
    5000 µg/g
  • Matrix:
    Mineral oil
  • Description:
    Chlorine - Cl @ 5000 µg/g in 75 cSt Mineral Oil
  • Size:
    500 ml
  • Cat. No.:
    77693-064