Order Entry
Puerto Rico
ContactUsLinkComponent
 
Chlorine Blank Standard (0 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
  77693-071
New Product
 :  
undefined
Chlorine Blank Standard (0 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
  77693-071
 :  VHG-CLOIL-BLK-100
New Product
 :  

 

  • Concentration:
    blank
  • Matrix:
    Mineral oil
  • Description:
    Chlorine Blank (0 wt%) in 75 cSt Mineral Oil
  • Size:
    100 ml
  • Cat. No.:
    77693-071