Sulfate Standard: SO4(2-) @ 100 µg/mL in H2O, VHG, LGC Standards
Catalog # 77695-333
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/mL
- Matrix:H₂O
- Description:Sulfate Standard: SO4(-2) @ 100 µg/mL in H2O
- Size:500 ml