Multi-Element 18 Components 200.8 Calibration Standard A2: 10 µg/mL Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Th, Tl, U, V, Zn; 50 µg/mL Se in 5% HNO3, tr. Tartaric Acid, VHG, LGC Standards
77693-148
New Product
:
- Concentration:50 µg/mL
- Matrix:5% HNO₃/tr. (CHOH)₂(COOH)₂
- Description:200.8 Calibration Standard A2: Al, As, Be, Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Sb, Th, Tl, U, V, Zn @ 10; Se @ 50 µg/mL in 5% HNO3, tr. Tartaric Acid
- Size:100 ml
- Cat. No.:77693-148