6020A & CLP-M ICS Analytes B: Ag @ 5; As, Cd, Se, Zn @ 10; Co, Cr, Cu, Mn, Ni, V @ 20 µg/mL in 2% HNO3, VHG, LGC Standards
77693-158
New Product
:
- Matrix:2% HNO₃
- Description:6020A & CLP-M ICS Analytes B: Ag @ 5; As, Cd, Se, Zn @ 10; Co, Cr, Cu, Mn, Ni, V @ 20 µg/mL in 2% HNO3
- Size:100 ml
- Cat. No.:77693-158