Order Entry
Puerto Rico
ContactUsLinkComponent
 
Antimony Standard: Sb @ 10 µg/mL in 1% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Catalog # 77693-162
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Antimony Standard: Sb @ 10 µg/mL in 1% HNO3, tr. Tartaric Acid, VHG, LGC Standards
Catalog # 77693-162
Supplier:  LGC Standards
Supplier Number:  VHG-LSBWTN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    1% HNO₃ + (CHOH)₂(COOH)₂
  • Description:
    Antimony Standard: Sb @ 10 µg/mL in 1% HNO3, tr. Tartaric Acid
  • Size:
    100 ml
  • Cat. No.:
    77693-162

Specifications

About this item