Order Entry
Puerto Rico
ContactUsLinkComponent
Tin Standard: Sn @ 10 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Tin Standard: Sn @ 10 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77695-346
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Tin Standard: Sn @ 10 µg/mL in 5% HNO3, tr. HF, VHG, LGC Standards
Catalog # 77695-346
Supplier:  LGC Standards
Supplier Number:  VHG-LSNNF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    Tin Standard: Sn @ 10 µg/mL in 5% HNO3, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77695-346

Specifications

About this item