Order Entry
Puerto Rico
ContactUsLinkComponent
Terbium Standard: Tb @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Terbium Standard: Tb @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77695-347
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Terbium Standard: Tb @ 10 µg/mL in 2% HNO3, VHG, LGC Standards
Catalog # 77695-347
Supplier:  LGC Standards
Supplier Number:  VHG-LTBN-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    2% HNO₃
  • Description:
    Terbium Standard: Tb @ 10 µg/mL in 2% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-347

Specifications

About this item