Order Entry
Puerto Rico
ContactUsLinkComponent
Multi-Element Standard 5 Components, Tune & Resolution Solution 1 for Method 200.8 & ILM05.2: 10 µg/mL each of Be, Mg, Co, In, Pb in 5% HNO3, VHG, LGC Standards
Multi-Element Standard 5 Components, Tune & Resolution Solution 1 for Method 200.8 & ILM05.2: 10 µg/mL each of Be, Mg, Co, In, Pb in 5% HNO3, VHG, LGC Standards
Catalog # 77695-348
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Element Standard 5 Components, Tune & Resolution Solution 1 for Method 200.8 & ILM05.2: 10 µg/mL each of Be, Mg, Co, In, Pb in 5% HNO3, VHG, LGC Standards
Catalog # 77695-348
Supplier:  LGC Standards
Supplier Number:  VHG-LTS2008D-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/mL
  • Matrix:
    5% HNO₃
  • Description:
    Tune & Resolution Solution 1 for Method 200.8 & ILM05.2: Be, Mg, Co, In, Pb @ 10 µg/mL in 5% HNO3
  • Size:
    100 ml
  • Cat. No.:
    77695-348

Specifications

About this item