Order Entry
Puerto Rico
ContactUsLinkComponent
Multi-Element Standard 4 Components: 5000 µg/g of Ca; 1600 µg/g each of Mg, P, Zn in Hydrocarbon Oil, VHG, LGC Standards
Multi-Element Standard 4 Components: 5000 µg/g of Ca; 1600 µg/g each of Mg, P, Zn in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-166
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Element Standard 4 Components: 5000 µg/g of Ca; 1600 µg/g each of Mg, P, Zn in Hydrocarbon Oil, VHG, LGC Standards
Catalog # 77693-166
Supplier:  LGC Standards
Supplier Number:  VHG-MA4-400G
New Product
CAS Number:  

Specifications

  • Concentration:
    1600 µg/g
  • Matrix:
    oil
  • Description:
    Ca @ 5000µg/g; Mg, P, Zn @ 1600µg/g in Hydrocarbon Oil, 400g
  • Size:
    400 g
  • Cat. No.:
    77693-166

Specifications

About this item