Order Entry
Puerto Rico
ContactUsLinkComponent
Multi-Element Standard 9 Components: 2500 µg/g each of B, Ba, Ca, Mg, Mo, Na, P, Si, Zn in 75cSt oil, VHG, LGC Standards
Multi-Element Standard 9 Components: 2500 µg/g each of B, Ba, Ca, Mg, Mo, Na, P, Si, Zn in 75cSt oil, VHG, LGC Standards
Catalog # 77693-167
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Multi-Element Standard 9 Components: 2500 µg/g each of B, Ba, Ca, Mg, Mo, Na, P, Si, Zn in 75cSt oil, VHG, LGC Standards
Catalog # 77693-167
Supplier:  LGC Standards
Supplier Number:  VHG-MA9-2500-200G
New Product
CAS Number:  

Specifications

  • Concentration:
    2500 µg/g
  • Matrix:
    oil
  • Description:
    B, Ba, Ca, Mg, Mo, Na, P, Si, Zn @ 2500 µg/g in 75cSt oil
  • Size:
    200 g
  • Cat. No.:
    77693-167

Specifications

About this item