Order Entry
Puerto Rico
ContactUsLinkComponent
Barium Standard: Ba @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
Barium Standard: Ba @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
  77693-175
New Product
 :  
undefined
Barium Standard: Ba @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
  77693-175
 :  VHG-OBA-1000-50G
New Product
 :  

 

  • Concentration:
    1000 µg/g
  • Matrix:
    oil
  • Description:
    Barium Standard: Ba @ 1000 µg/g in Hydrocarbon Oil
  • Size:
    50 g
  • Cat. No.:
    77693-175