Order Entry
Puerto Rico
ContactUsLinkComponent
 
Tin Standard: Sn @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
  77695-370
New Product
 :  
undefined
Tin Standard: Sn @ 1000 µg/g in Hydrocarbon Oil, VHG, LGC Standards
  77695-370
 :  VHG-OSN-1000-50G
New Product
 :  

 

  • Concentration:
    1000 µg/g
  • Matrix:
    oil
  • Description:
    Tin Standard: Sn @ 1000 µg/g in Hydrocarbon Oil
  • Size:
    50 g
  • Cat. No.:
    77695-370