Order Entry
Puerto Rico
ContactUsLinkComponent
Antimony - Sb @ 1000 µg/mL in 5% HNO3/0.1% HF, VHG, LGC Standards
Antimony - Sb @ 1000 µg/mL in 5% HNO3/0.1% HF, VHG, LGC Standards
Catalog # 77693-253
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Antimony - Sb @ 1000 µg/mL in 5% HNO3/0.1% HF, VHG, LGC Standards
Catalog # 77693-253
Supplier:  LGC Standards
Supplier Number:  VHG-PSBNF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    Antimony - Sb @ 1000 µg/mL in 5% HNO3/0.1% HF
  • Size:
    100 ml
  • Cat. No.:
    77693-253

Specifications

About this item