Sulfur @ 10 µg/g, from Polysulfide Oil, VHG, LGC Standards
Catalog # 77695-387
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:10 µg/g
- Description:Sulfur @ 10 µg/g, from Polysulfide Oil
- Size:500 ml
- Cat. No.:77695-387