Order Entry
Puerto Rico
ContactUsLinkComponent
Tellurium Standard: Te @ 1000 µg/mL in 30% HCl, VHG, LGC Standards
Tellurium Standard: Te @ 1000 µg/mL in 30% HCl, VHG, LGC Standards
Catalog # 77695-402
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Tellurium Standard: Te @ 1000 µg/mL in 30% HCl, VHG, LGC Standards
Catalog # 77695-402
Supplier:  LGC Standards
Supplier Number:  VHG-PTEH-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    30% HCl
  • Description:
    Tellurium Standard: Te @ 1000 µg/mL in 30% HCl
  • Size:
    100 ml
  • Cat. No.:
    77695-402

Specifications

About this item