Order Entry
Puerto Rico
ContactUsLinkComponent
Titanium Standard: Ti @ 1000 µg/mL in H2O, tr. F-, VHG, LGC Standards
Titanium Standard: Ti @ 1000 µg/mL in H2O, tr. F-, VHG, LGC Standards
Catalog # 77695-408
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Titanium Standard: Ti @ 1000 µg/mL in H2O, tr. F-, VHG, LGC Standards
Catalog # 77695-408
Supplier:  LGC Standards
Supplier Number:  VHG-PTIW-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 µg/mL
  • Matrix:
    5% HNO₃/tr HF
  • Description:
    Titanium Standard: Ti @ 1000 µg/mL in H2O, tr. HF
  • Size:
    100 ml
  • Cat. No.:
    77695-408

Specifications

About this item