Sulfur Standard: S @ 250 µg/g (0.0250 wt%) in 20 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-446
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:250 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 250 µg/g (0.0250 wt%) in 20 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-446