Sulfur Standard: S @ 100 µg/g in B100 Biodiesel, VHG, LGC Standards
Catalog # 77695-463
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg/g
- Description:Sulfur Standard: S @ 100 µg/g in B100 Biodiesel
- Size:100 ml
- Cat. No.:77695-463