Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfur & Chlorine Standard: S, Cl @ 5000 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
Sulfur & Chlorine Standard: S, Cl @ 5000 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-473
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Sulfur & Chlorine Standard: S, Cl @ 5000 µg/g in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-473
Supplier:  LGC Standards
Supplier Number:  VHG-SCLOIL-5000-500
New Product
CAS Number:  

Specifications

  • Concentration:
    5000 µg/g
  • Matrix:
    Mineral oil
  • Description:
    Sulfur & Chlorine Standard: S, Cl @ 5000 µg/g in 75 cSt Mineral Oil
  • Size:
    500 ml
  • Cat. No.:
    77695-473

Specifications

About this item