Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in #2 Diesel Fuel, VHG, LGC Standards
77695-477
New Product
:
- Concentration:1000 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in #2 Diesel Fuel
- Size:100 ml
- Cat. No.:77695-477