Sulfur Standard: S @ 25 µg/g (0.0025 wt%) in #2 Diesel Fuel, VHG, LGC Standards
Catalog # 77695-488
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:25 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 25 µg/g (0.0025 wt%) in #2 Diesel Fuel
- Size:100 ml
- Cat. No.:77695-488