Sulfur Standard: S @ 250 µg/g (0.0250 wt%) in Diesel, VHG, LGC Standards
Catalog # 77695-487
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:250 µg
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 250 µg/g (0.0250 wt%) in Diesel
- Size:100 ml
- Cat. No.:77695-487