Sulfur Standard: S @ 20000 µg/g (2.00 wt%) in #2 Diesel Fuel, VHG, LGC Standards
77695-489
New Product
:
- Concentration:20000 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 20000 µg/g (2.00 wt%) in #2 Diesel Fuel
- Size:100 g
- Cat. No.:77695-489