Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in #2 Diesel Fuel, VHG, LGC Standards
Catalog # 77695-490
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:3000 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in #2 Diesel Fuel
- Size:100 ml
- Cat. No.:77695-490