Sulfur Standard: S @ 400 µg/g (0.0400 wt%) in #2 Diesel Fuel, VHG, LGC Standards
Catalog # 77695-493
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:400 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 400 µg/g (0.0400 wt%) in #2 Diesel Fuel
- Size:100 ml
- Cat. No.:77695-493