Sulfur Standard: S @ 750 µg/g (0.0750 wt%) in #2 Diesel Fuel, VHG, LGC Standards
77695-502
New Product
:
- Concentration:750 µg/g
- Matrix:#2 diesel fuel
- Description:Sulfur Standard: S @ 750 µg/g (0.0750 wt%) in #2 Diesel Fuel
- Size:100 ml
- Cat. No.:77695-502