Sulfur Standard: S @ 100 µg/g (0.0100 wt%) in Isooctane, VHG, LGC Standards
Catalog # 77695-508
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:100 µg
- Description:Sulfur Standard: S @ 100 µg/g (0.0100 wt%) in Isooctane
- Size:100 ml
- Cat. No.:77695-508