Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in Isooctane, VHG, LGC Standards
Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in Isooctane, VHG, LGC Standards
Catalog # 77695-516
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in Isooctane, VHG, LGC Standards
Catalog # 77695-516
Supplier:  LGC Standards
Supplier Number:  VHG-SISO-3000-100
New Product
CAS Number:  

Specifications

  • Concentration:
    3000 µg
  • Description:
    Sulfur Standard: S @ 3000 µg/g (0.300 wt%) in Isooctane
  • Size:
    100 ml
  • Cat. No.:
    77695-516

Specifications

About this item