Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene, VHG, LGC Standards
Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene, VHG, LGC Standards
Catalog # 77695-536
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene, VHG, LGC Standards
Catalog # 77695-536
Supplier:  LGC Standards
Supplier Number:  VHG-SISO80TOL20-1000-100
New Product
CAS Number:  

Specifications

  • Concentration:
    1000 ug/g
  • Description:
    Sulfur Standard: S @ 1000 ug/g (0.1 wt%) in 80% Isooctane/20% Toluene
  • Size:
    100 ml
  • Cat. No.:
    77695-536

Specifications

About this item